Palladium(II) chloride

Warning

Synonyms: Palladium dichloride

76050-5G 76050-1G 76050-25G
220095-346EA 517.15 USD
220095-346 220095-344 220095-345
Palladium(II) chloride
Palladium(II) chloride
Formula: PdCl₂
MW: 177.33 g/mol
Melting Pt: 678 °C
Density: 4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8,III
Merck Index: 13,07058

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR