Palladium 5% on barium sulfate

76022-10G 76022-50G
220095-326EA 186.92 USD
220095-326 220095-327
Palladium 5% on barium sulfate
Palladium
Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now

Specification Test Results

Assay 5% Pd Basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR