4,6-Dimethoxyindole

75065-5G-F 75065-1G-F
220095-746EA 104.66 USD
220095-746 220095-745
4,6-Dimethoxyindole
4,6-Dimethoxyindole
Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
MDL Number: MFCD00128213
CAS Number: 23659-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR