1-Butyl-3-methylimidazolium octylsulfate

75059-5G-F 75059-50G-F
220095-743EA 83.5 USD
220095-743 220095-742
1-Butyl-3-methylimidazolium octylsulfate
1-Butyl-3-methylimidazolium octylsulphate
Formula: C₁₆H₃₂N₂O₄S
MW: 348.51 g/mol
MDL Number: MFCD03788914
CAS Number: 445473-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR