1-Octanesulfonic acid sodium salt monohydrate ≥97.0% (by titrimetric analysis)

Synonyms: Sodium 1-octansulphonate monohydrate, Sodium 1-octanesulphonate monohydrate

74885-50G 74885-10G
220095-715EA 305.85 USD
220095-715 220095-714
1-Octanesulfonic acid sodium salt monohydrate ≥97.0% (by titrimetric analysis)
1-Octanesulphonic acid sodium salt monohydrate
Formula: C₈H₁₇NaO₃S·1H₂O
MW: 234.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 207596-29-0
EINECS: 226-195-4

Order Now

Specification Test Results

Assay 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR