(±)-2-Octanol ≥99.5% (by GC)

Danger

Synonyms: (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol

74858-100ML 74858-25ML
220095-705EA 501.85 USD
220095-705 220095-706
(±)-2-Octanol ≥99.5% (by GC)
(±)-2-Octanol
Formula: C₈H₁₈O
MW: 130.23 g/mol
Boiling Pt: 180 °C (1013 hPa)
Melting Pt: –39 °C
Density: 0.818 g/cm³ (20 °C)
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00004591
CAS Number: 123-96-6
EINECS: 204-667-0
UN: 1987
ADR: 3,III
Merck Index: 13,06785

Order Now

Specification Test Results

Assay 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR