1-Octadecene

74740-100ML 74740-500ML
220095-687EA 156.15 USD
220095-687 220095-688
1-Octadecene
1-Octadecene
Formula: C₁₈H₃₆
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 17.5 °C
Density: 0.789 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009003
CAS Number: 112-88-9
EINECS: 204-012-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR