Benzyl nicotinate

Warning

Synonyms: 3-Picolinic acid benzyl ester, Pyridine-3-carboxylic acid benzyl ester, Benzyl 3-pyridinecarboxylate, 3-Pyridinecarboxylic acid benzyl ester, Nicotinic acid benzyl ester

72351-100ML 72351-500ML
220094-442EA 133.54 USD
220094-442 220094-443
Benzyl nicotinate
Benzyl nicotinate
Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
Boiling Pt: 179…180 °C (8 hPa)
Melting Pt: 24 °C
Density: 1.17 g/cm³ (20 °C)
MDL Number: MFCD00023584
CAS Number: 94-44-0
EINECS: 202-332-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR