Nickel(II) bromide trihydrate

Danger

72243-1KG 72243-250G
220094-398EA 830.77 USD
220094-398 220094-399
Nickel(II) bromide trihydrate
Nickel(II) bromide trihydrate
Formula: Br₂Ni·3H₂O
MW: 272.54 g/mol
Melting Pt: 962 - 964 °C
MDL Number: MFCD00011141
CAS Number: 7789-49-3
EINECS: 236-665-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98.0%
(AT)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR