4-Bromo-2-fluoropyridine

Warning

722421-5G 722421-1G
220094-397EA 156.15 USD
220094-397 220094-396
4-Bromo-2-fluoropyridine
4-Bromo-2-fluoropyridine
Formula: C₅H₃BrFN
MW: 175.99 g/mol
MDL Number: MFCD04112504
CAS Number: 128071-98-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR