2-Amino-3-bromo-5-fluoropyridine

722383-1G 722383-5G
220094-388EA 63.54 USD
220094-388 220094-389
2-Amino-3-bromo-5-fluoropyridine
2-Amino-3-bromo-5-fluoropyridine
Formula: C₅H₄BrFN₂
MDL Number: MFCD03092924
CAS Number: 869557-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR