2,3-Difluoropyridine

Danger

718173-5G 718173-1G
220093-822EA 119.54 USD
220093-822 220093-821
2,3-Difluoropyridine
2,3-Difluoropyridine
Formula: C₅H₃F₂N
MW: 115.08 g/mol
MDL Number: MFCD04114143
CAS Number: 1513-66-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR