3-Iodo-4-methoxybenzoic acid ≥97%

Synonyms: 3-Iodo-p-anisic acid

717800-25G 717800-5G
220093-784EA 230.46 USD
220093-784 220093-785
3-Iodo-4-methoxybenzoic acid ≥97%
3-Iodo-4-methoxybenzoic acid
Formula: C₈H₇IO₃
MW: 278.05 g/mol
MDL Number: MFCD00017347
CAS Number: 68507-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR