Disodium metasilicate pentahydrate ≥95.0% (by titrimetric analysis)

Danger

Synonyms: Sodium metasilicate pentahydrate

71746-1KG 71746-250G
220093-717EA 45.75 USD
220093-717 220093-718
Disodium metasilicate pentahydrate ≥95.0% (by titrimetric analysis)
Disodium metasilicate pentahydrate
Formula: Na₂SiO₃·5H₂O
MW: 212.14 g/mol
Melting Pt: 72.2 °C
Density: 1.749 g/cm³ (20 °C)
MDL Number: MFCD00149176
CAS Number: 10213-79-3
EINECS: 229-912-9
UN: 3253
ADR: 8,III
Merck Index: 13,08716

Order Now

Specification Test Results

Assay 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR