DL-Sodium lactate ≥99%

Synonyms: (±)-Lactic acid sodium salt, (±)-2-Hydroxypropionic acid sodium salt, Lactate sodium, DL-Lactic acid sodium salt, (±)-2-Hydroxypropanoic acid sodium salt, Sodium 2-hydroxypropanoate

71720-25G 71720-5G
220093-676EA 276.15 USD
220093-676 220093-677
DL-Sodium lactate ≥99%
DL-Sodium lactate
Formula: H₃CCH(OH)COONa
MW: 112.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065400
CAS Number: 72-17-3
EINECS: 200-772-0
Merck Index: 13,08709

Order Now

Specification Test Results

Assay 99% (NT)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR