L(+)-Sodium lactate ≥99%

Synonyms: Sodium (S)-2-hydroxypropanoate, (S)-(+)-2-Hydroxypropanoic acid sodium salt, (S)-(+)-2-Hydroxypropionic acid sodium salt, Sodium-(S)-lactate, (S)-(+)-Lactic acid sodium salt, L(+)-Lactic acid sodium salt

71718-10G 71718-50G
220093-669EA 114.15 USD
220093-669 220093-670
L(+)-Sodium lactate ≥99%
L(+)-Sodium lactate
Formula: C₃H₅NaO₃
MW: 112.06 g/mol
MDL Number: MFCD00066576
CAS Number: 867-56-1
EINECS: 212-762-3

Order Now

Specification Test Results

Assay 99% (NT)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR