2-Chloro-3-iodopyridine

Warning

716030-1G 716030-5G
220091-706EA 103.95 USD
220091-706 220091-707
2-Chloro-3-iodopyridine
2-Chloro-3-iodopyridine
Formula: C₅H₃ClIN
MW: 239.44 g/mol
MDL Number: MFCD00661298
CAS Number: 78607-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR