4-Chloro-3-fluoropyridine

Danger

71585-25G-F 71585-5G-F
220093-372EA 398.85 USD
220093-372 220093-373
4-Chloro-3-fluoropyridine
4-Chloro-3-fluoropyridine
Formula: C₅H₃ClFN
MW: 131.54 g/mol
MDL Number: MFCD03453233
CAS Number: 2546-56-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR