3-Phenylbenzoic acid

Synonyms: Biphenyl-3-carboxylic acid, 3-Biphenylcarboxylic acid, 1,1'-Biphenyl-3-carboxylic acid, [1,1'-Biphenyl]-3-carboxylic acid

715204-1G 715204-5G
220093-291EA 131.54 USD
220093-291 220093-292
3-Phenylbenzoic acid
3-Phenylbenzoic acid
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
MDL Number: MFCD00045846
CAS Number: 716-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR