3-Hydroxy-5-nitrobenzoic acid ≥97%

714402-1G 714402-5G
220093-228EA 94.23 USD
220093-228 220093-229
3-Hydroxy-5-nitrobenzoic acid ≥97%
3-Hydroxy-5-nitrobenzoic acid
Formula: C₇H₅NO₅
MW: 183.12 g/mol
MDL Number: MFCD01098212
CAS Number: 78238-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR