1-Ethyl-3-methylimidazolium iodide

Synonyms: 1-ethyl-3-methylimidazolium iodide

713031-50G 713031-5G
220092-865EA 205 USD
220092-865 220092-866
1-Ethyl-3-methylimidazolium iodide
1-Ethyl-3-methylimidazolium iodide
Formula: C₆H₁₁IN₂
MW: 238.07 g/mol
MDL Number: MFCD03701101
CAS Number: 35935-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR