1-Ethyl-2,3-dimethylimidazoliumtetrafluoroborate

Synonyms: 1-ethyl-2,3-dimethylimidazolium tetrafluoroborate

713023-5G 713023-50G
220092-864EA 218.4 USD
220092-864 220092-863
1-Ethyl-2,3-dimethylimidazoliumtetrafluoroborate
1-Ethyl-2,3-dimethylimidazoliumtetrafluoroborate
Formula: C₇H₁₃BF₄N₂
MW: 212 g/mol
MDL Number: MFCD08458929
CAS Number: 307492-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR