1-Ethyl-2,3-dimethylimidazoliumhexafluorophosphate

Synonyms: 1-Ethyl-2,3-dimethyl-1H-imidazol-3-ium hexafluorophosphate(V)

713015-50G 713015-5G
220092-861EA 218.4 USD
220092-861 220092-862
1-Ethyl-2,3-dimethylimidazoliumhexafluorophosphate
1-Ethyl-2,3-dimethylimidazoliumhexafluorophosphate
Formula: C₇H₁₃F₆N₂P
MW: 270.16 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Flash Pt: >100 °C
MDL Number: MFCD08458503
CAS Number: 292140-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR