(R)-(+)-1-(2-Naphthyl)ethylamine

Synonyms: (R)-1-(2-Naphthyl)ethylamine

70940-1G 70940-5G
220092-715EA 7349.23 USD
220092-715 220092-716
(R)-(+)-1-(2-Naphthyl)ethylamine
(R)-(+)-1-(2-Naphthyl)ethylamine
Formula: C₁₂H₁₃N
MW: 171.24 g/mol
MDL Number: MFCD00085367
CAS Number: 3906-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR