Iron diarsenide

Danger

709271-5G 709271-1G
220092-698EA 747.69 USD
220092-698 220092-697
Iron diarsenide
Iron diarsenide
Formula: As₂Fe
MW: 205.69 g/mol
CAS Number: 12006-21-2
EINECS: 234-485-7
UN: 1557
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR