Trizinc diarsenide

Danger

Synonyms: Zinc arsenide, Trizinc diarsenide

709247-1G 709247-5G
220092-695EA 286.15 USD
220092-695 220092-696
Trizinc diarsenide
Trizinc diarsenide
Formula: As₂Zn₃
MW: 346.01 g/mol
MDL Number: MFCD00049629
CAS Number: 12006-40-5
EINECS: 234-486-2
UN: 1557
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR