2-Methoxyethyl isothiocyanate ≥95%

709123-1G 709123-5G
220092-678EA 46.31 USD
220092-678 220092-679
2-Methoxyethyl isothiocyanate ≥95%
2-Methoxyethyl isothiocyanate
Formula: C₄H₇NOS
MW: 117.17 g/mol
Boiling Pt: 88.5…92 °C (23.5 mmHg)
Density: 1.08 g/cm³ (20 °C)
MDL Number: MFCD00041126
CAS Number: 38663-85-3
EINECS: 254-070-4

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR