1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate

Synonyms: 1-Butyl-2,3-dimethylimidazolium Tetrafluoroborate, [BDiMIM] [BF4]

70863-50G-F 70863-5G-F
220092-620EA 297 USD
220092-620 220092-621
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate
Formula: C₉H₁₇BF₄N₂
MW: 240.05 g/mol
Density: 1.2 g/cm³ (20 °C)
MDL Number: MFCD03427618
CAS Number: 402846-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR