2-Amino-4-bromophenol

Synonyms: 5-Bromo-2-hydroxyaniline

708356-1G 708356-5G
220092-521EA 63.97 USD
220092-521 220092-522
2-Amino-4-bromophenol
2-Amino-4-bromophenol
Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD00235171
CAS Number: 40925-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR