3-Chloro-p-cresol ≥97%

Synonyms: 3-Chloro-4-methylphenol, 1-Chloro-3-hydroxy-6-methylbenzene

708224-5G 708224-1G
220092-509EA 36.72 USD
220092-509 220092-508
3-Chloro-p-cresol ≥97%
3-Chloro-p-cresol
Formula: C₇H₇ClO
MW: 142.58 g/mol
MDL Number: MFCD00060319
CAS Number: 615-62-3
EINECS: 210-439-1
UN: 3437
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR