Methyl-3,4-diaminobenzoate ≥97%

Synonyms: 3,4-Diamino benzoate, 3,4-Diaminobenzoic acid methyl ester, Methyl 3,4-diaminobenzoate

705985-5G 705985-25G
220092-260EA 189.54 USD
220092-260 220092-259
Methyl-3,4-diaminobenzoate ≥97%
Methyl-3,4-diaminobenzoate
Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 108…110 °C
MDL Number: MFCD00017098
CAS Number: 36692-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR