2-(N,N-Dimethylamino)phenylboronic acid pinacol ester

Synonyms: N,N-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

704652-5G 704652-1G
220092-108EA 32.95 USD
220092-108 220092-107
2-(N,N-Dimethylamino)phenylboronic acid pinacol ester
2-(N,N-Dimethylamino)phenylboronic acid pinacol ester
Formula: C₁₄H₂₂BNO₂
MW: 247.14 g/mol
MDL Number: MFCD05864304
CAS Number: 832114-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR