Strontium acetylacetonate hydrate

Synonyms: Bis(2,4-pentanedionato-O,O')strontium hydrate, Strontium bis(2,4-pentanedionato-O,O') hydrate, Strontium 2,4-pentanedionate hydrate

704512-5G 704512-25G
220092-085EA 210.77 USD
220092-085 220092-084
Strontium acetylacetonate hydrate
Strontium acetylacetonate hydrate
Formula: C₁₀H₁₄O₄Sr·H₂O
MW: 303.85 g/mol
MDL Number: MFCD00013502
CAS Number: 12193-47-4
EINECS: 235-366-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR