6-Fluoropyridine-2-carbonitrile

Synonyms: 6-Fluoropicolinonitrile, 2-Cyano-6-fluoropyridine

704024-1G 704024-250MG
220091-199EA 140 USD
220091-199 220091-200
6-Fluoropyridine-2-carbonitrile
6-Fluoropyridine-2-carbonitrile
Formula: C₆H₃FN₂
MW: 122.1 g/mol
MDL Number: MFCD09263323
CAS Number: 3939-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR