5-Fluoropyridine-2-carbonitrile

Synonyms: 5-Fluoro-2-pyridinecarbonitrile, 2-Cyano-5-fluoropyridine

703982-1G 703982-5G
220091-190EA 35.97 USD
220091-190 220091-191
5-Fluoropyridine-2-carbonitrile
5-Fluoropyridine-2-carbonitrile
Formula: C₆H₃FN₂
MW: 122.1 g/mol
MDL Number: MFCD08741371
CAS Number: 327056-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR