2-Chloro-4-fluorobenzoyl chloride ≥97%

Synonyms: 2-Chloro-4-fluorobenzene-1-carbonyl chloride, 2-Chloro-4-fluorobenzoylchloride

703958-25G 703958-5G
220092-018EA 227.23 USD
220092-018 220092-019
2-Chloro-4-fluorobenzoyl chloride ≥97%
2-Chloro-4-fluorobenzoyl chloride
Formula: C₇H₃Cl₂FO
MW: 193 g/mol
MDL Number: MFCD00084954
CAS Number: 21900-54-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR