3-Bromo-5-nitrobenzaldehyde ≥97%

702986-250MG 702986-1G
220091-668EA 74.85 USD
220091-668 220091-667
3-Bromo-5-nitrobenzaldehyde ≥97%
3-Bromo-5-nitrobenzaldehyde
Formula: C₇H₄BrNO₃
MW: 230.02 g/mol
MDL Number: MFCD00100097
CAS Number: 355134-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR