3-Bromo-4-hydroxybenzoic acid ≥97%

699926-5G 699926-25G
220091-303EA 103.17 USD
220091-303 220091-302
3-Bromo-4-hydroxybenzoic acid ≥97%
3-Bromo-4-hydroxybenzoic acid
Formula: C₇H₅BrO₃
MW: 217.02 g/mol
MDL Number: MFCD00017547
CAS Number: 14348-41-5
EINECS: 238-299-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR