3-Iodophenylboronic acid pinacol ester ≥97%

Synonyms: 3-Iodobenzeneboronic acid pinacol ester, 2-(3-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

699462-5G 699462-1G
220090-793EA 105.92 USD
220090-793 220090-792
3-Iodophenylboronic acid pinacol ester ≥97%
3-Iodophenylboronic acid pinacol ester
Formula: C₁₂H₁₆BIO₂
MW: 329.97 g/mol
MDL Number: MFCD09953498
CAS Number: 408492-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR