7-Methylxanthine

Synonyms: 2,6-Dihydroxy-7-methylpurine, 7-Methyl-3,7-dihydro-1H-purine-2,6-dione, 3,7-Dihydro-7-methyl-1H-purine-2,6-dione

69723-250MG 69723-1G
220090-959EA 227.31 USD
220090-959 220090-958
7-Methylxanthine
7-Methylxanthine
Formula: C₆H₆N₄O₂
MW: 166.14 g/mol
MDL Number: MFCD00037979
CAS Number: 552-62-5
EINECS: 209-019-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR