2-Bromo-p-toluonitrile ≥97%

697206-25G 697206-5G
220090-954EA 118.46 USD
220090-954 220090-955
2-Bromo-p-toluonitrile ≥97%
2-Bromo-p-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD00158968
CAS Number: 42872-73-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR