1-Methylxanthine

Synonyms: 3,7-Dihydro-1-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-1-methylpurine, 1-Methyl-1H-purine-2,6(3H,7H)-dione

69720-250MG 69720-1G
220090-953EA 317.31 USD
220090-953 220090-952
1-Methylxanthine
1-Methylxanthine
Formula: C₆H₆N₄O₂
MW: 166.14 g/mol
MDL Number: MFCD00005561
CAS Number: 6136-37-4
EINECS: 228-108-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR