2-Bromo-m-toluonitrile ≥97%

697176-1G 697176-5G
220089-888EA 43.08 USD
220089-888 220089-889
2-Bromo-m-toluonitrile ≥97%
2-Bromo-m-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD09025664
CAS Number: 263159-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR