3-Amino-4-methylpyridine

697141-5G 697141-25G
220089-886EA 81.31 USD
220089-886 220089-885
3-Amino-4-methylpyridine
3-Amino-4-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00128871
CAS Number: 3430-27-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR