3-Fluoropyridine-2-carbonitrile

Synonyms: 3-Fluoro-2-pyridinecarbonitrile, 2-Cyano-3-fluoropyridine, 3-Fluoropicolinonitrile

697109-1G 697109-5G
220090-946EA 30.69 USD
220090-946 220090-947
3-Fluoropyridine-2-carbonitrile
3-Fluoropyridine-2-carbonitrile
Formula: C₆H₃FN₂
MW: 122.1 g/mol
MDL Number: MFCD06797501
CAS Number: 97509-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR