2-Fluoro-3-pyridineboronic acid

Synonyms: 2-Fluoro-3-pyridinylboronic acid, (2-Fluoropyridin-3-yl)boronic acid, 2-Fluoropyridine-3-boronic acid

696579-1G 696579-5G
220090-895EA 121.15 USD
220090-895 220090-896
2-Fluoro-3-pyridineboronic acid
2-Fluoro-3-pyridineboronic acid
Formula: C₅H₅BFNO₂
MW: 140.91 g/mol
MDL Number: MFCD03411557
CAS Number: 174669-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR