3-Chloro-2-methoxybenzoic acid ≥97%

696382-5G 696382-1G
220090-886EA 122.77 USD
220090-886 220090-885
3-Chloro-2-methoxybenzoic acid ≥97%
3-Chloro-2-methoxybenzoic acid
Formula: C₈H₇ClO₃
MW: 186.59 g/mol
MDL Number: MFCD00179306
CAS Number: 3260-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR