3-Chloro-5-methoxybenzoic acid ≥97%

696374-5G 696374-1G
220090-884EA 156.15 USD
220090-884 220090-883
3-Chloro-5-methoxybenzoic acid ≥97%
3-Chloro-5-methoxybenzoic acid
Formula: C₈H₇ClO₃
MW: 186.59 g/mol
MDL Number: MFCD01317994
CAS Number: 82477-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR