5-Bromo-2-methoxybenzoic acid ≥97%

694789-1G 694789-5G
220090-224EA 32.2 USD
220090-224 220090-225
5-Bromo-2-methoxybenzoic acid ≥97%
5-Bromo-2-methoxybenzoic acid
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
MDL Number: MFCD00093713
CAS Number: 2476-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR