2-Amino-3-iodo-5-nitropyridine

Synonyms: 3-Iodo-5-nitropyridin-2-amine, 3-Iodo-5-nitro-2-pyridylamine

694738-1G 694738-5G
220086-992EA 66 USD
220086-992 220086-993
2-Amino-3-iodo-5-nitropyridine
2-Amino-3-iodo-5-nitropyridine
Formula: C₅H₄IN₃O₂
MW: 265.01 g/mol
MDL Number: MFCD07368863
CAS Number: 25391-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR